ENA371177450 Enamine

3-(2,2,2-trifluoroethyl)azetidine hydrochloride

  • MDL number MFCD22375347
  • Cas Number : 1394041-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107382
Availability details for
Quantity
  • Availability
  • Add to Cart