ENA371177452 Enamine

octahydro-1-benzofuran-4-amine

  • MDL number MFCD22375352
  • Cas Number : 1394040-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107487
Availability details for
Quantity
  • Availability
  • Add to Cart