ENA371177453 Enamine

2-[(4-methoxyphenyl)methyl]azetidine

  • MDL number MFCD22375353
  • Cas Number : 1394042-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107497
Availability details for
Quantity
  • Availability
  • Add to Cart