ENA371177454 Enamine

5-chloro-3-methyl-1-(2-methylpropyl)-4-(pyrrolidin-2-yl)-1H-pyrazole

  • MDL number MFCD22375355
  • Cas Number : 1394041-83-8
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-1-isobutyl-3-methyl-4-(2-pyrrolidinyl)-1H-pyrazole
Inchi Code 1S/C12H20ClN3/c1-8(2)7-16-12(13)11(9(3)15-16)10-5-4-6-14-10/h8,10,14H,4-7H2,1-3H3
Inchi Key HZRKIQZSTBBORH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-107548

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-107548
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart