ENA371177455 Enamine

1-(5-bromothiophen-2-yl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD22375360
  • Cas Number : 1394040-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107558
Availability details for
Quantity
  • Availability
  • Add to Cart