ENA371177459 Enamine

2-(5-methyl-2H-1,2,3,4-tetrazol-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22375365
  • Cas Number : 1369803-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107740
Availability details for
Quantity
  • Availability
  • Add to Cart