ENA371177462 Enamine

1-[3-(4-ethylphenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD22375368
  • Cas Number : 1394042-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192...194
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107744
Availability details for
Quantity
  • Availability
  • Add to Cart