ENA371177463 Enamine

1-[3-(4-fluoro-3-methylphenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD22375369
  • Cas Number : 1394041-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107745

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107745
Availability details for
Quantity
  • Availability
  • Add to Cart