ENA371177464 Enamine

tert-butyl N-(4-methyl-1,3-thiazol-5-yl)carbamate

  • MDL number MFCD22375371
  • Cas Number : 1318758-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107782
Availability details for
Quantity
  • Availability
  • Add to Cart