ENA371177477 Enamine

2-(oxan-4-yloxy)pyridine-4-carboximidamide hydrochloride

  • MDL number MFCD22375388
  • Cas Number : 1394041-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108018
Availability details for
Quantity
  • Availability
  • Add to Cart