ENA371177478 Enamine

tert-butyl 4-(4-ethyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-3-yl)piperidine-1-carboxylate

  • MDL number MFCD22375389
  • Cas Number : 1394042-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108021
Availability details for
Quantity
  • Availability
  • Add to Cart