ENA371177479 Enamine

2-(benzylamino)-1-[3-(benzyloxy)phenyl]ethan-1-ol

  • MDL number MFCD22375390
  • Cas Number : 1268868-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108023
Availability details for
Quantity
  • Availability
  • Add to Cart