ENA371177481 Enamine

1-(4-benzylmorpholin-2-yl)propan-1-amine

  • MDL number MFCD22375392
  • Cas Number : 1394040-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108030
Availability details for
Quantity
  • Availability
  • Add to Cart