ENA371177485 Enamine

1-[3-(3-fluoro-4-methylphenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD22375396
  • Cas Number : 1394041-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108082

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108082
Availability details for
Quantity
  • Availability
  • Add to Cart