ENA371177493 Enamine

4-[(3-methoxyphenyl)methylidene]piperidine hydrochloride

  • MDL number MFCD22375405
  • Cas Number : 584555-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108245

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108245
Availability details for
Quantity
  • Availability
  • Add to Cart