ENA371177494 Enamine

3-[(ethylamino)methyl]benzamide hydrochloride

  • MDL number MFCD22375406
  • Cas Number : 1394040-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108247
Availability details for
Quantity
  • Availability
  • Add to Cart