ENA371177497 Enamine

(6-fluoro-1,3-benzoxazol-2-yl)methanol

  • MDL number MFCD22375409
  • Cas Number : 1394041-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108250
Availability details for
Quantity
  • Availability
  • Add to Cart