ENA371177499 Enamine

2-(2-aminoethoxy)acetamide hydrochloride

  • MDL number MFCD22375411
  • Cas Number : 1394042-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108305

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108305
Availability details for
Quantity
  • Availability
  • Add to Cart