ENA371177506 Enamine

5-methyl-4-[(methylsulfanyl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD22375418
  • Cas Number : 1394041-05-4
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108347
Availability details for
Quantity
  • Availability
  • Add to Cart