ENA371177508 Enamine

2-amino-4-(ethanesulfonyl)benzoic acid

  • MDL number MFCD22375420
  • Cas Number : 949033-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108360
Availability details for
Quantity
  • Availability
  • Add to Cart