ENA371177533 Enamine

5-(chlorodifluoromethyl)-1-ethyl-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD22375434
  • Cas Number : 1394042-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90928
Availability details for
Quantity
  • Availability
  • Add to Cart