ENA371177538 Enamine

methyl 4,6-dichloropyrimidine-5-carboxylate

  • MDL number MFCD10698883
  • Cas Number : 87600-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-49
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-106517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106517
Availability details for
Quantity
  • Availability
  • Add to Cart