ENA371177541 Enamine

6,6-dimethyl-3-[(morpholin-4-yl)carbonyl]-4,5,6,7-tetrahydro-1-benzothiophen-2-amine

  • MDL number MFCD22375256
  • Cas Number : 1384594-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106521
Availability details for
Quantity
  • Availability
  • Add to Cart