ENA371177542 Enamine

[(pyridin-2-yl)methyl]({[3-(trifluoromethyl)phenyl]methyl})amine

  • MDL number MFCD12447218
  • Cas Number : 1019564-99-8
  • Molecular Weight : 266.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106523
Availability details for
Quantity
  • Availability
  • Add to Cart