ENA371177545 Enamine

4,6-dichloro-N-(2-methoxyethyl)-1,3,5-triazin-2-amine

  • MDL number MFCD17226234
  • Cas Number : 13807-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106529

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106529
Availability details for
Quantity
  • Availability
  • Add to Cart