ENA371177546 Enamine

(2R)-2-(carbamoylamino)pentanoic acid

  • MDL number MFCD20656930
  • Cas Number : 211681-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102901
Availability details for
Quantity
  • Availability
  • Add to Cart