ENA371177552 Enamine

3-amino-1-phenylbutan-1-ol

  • MDL number MFCD20690972
  • Cas Number : 25756-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106539
Availability details for
Quantity
  • Availability
  • Add to Cart