ENA371177562 Enamine

3-(2-methylpropyl)piperidine-2,6-dione

  • MDL number MFCD12405589
  • Cas Number : 1341348-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106549
Availability details for
Quantity
  • Availability
  • Add to Cart