ENA371177566 Enamine

6-(3-chlorophenoxy)pyridine-2-carboxylic acid

  • MDL number MFCD17455714
  • Cas Number : 137640-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106553

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106553
Availability details for
Quantity
  • Availability
  • Add to Cart