ENA371177576 Enamine

[5-(propan-2-yl)-1,3,4-thiadiazol-2-yl]methanamine hydrochloride

  • MDL number MFCD11841159
  • Cas Number : 1228880-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103534

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103534
Availability details for
Quantity
  • Availability
  • Add to Cart