ENA371177602 Enamine

5-fluoro-1-benzothiophene

  • MDL number MFCD11110411
  • Cas Number : 70060-12-7
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-1-benzothiophene
Inchi Code 1S/C8H5FS/c9-7-1-2-8-6(5-7)3-4-10-8/h1-5H
Inchi Key VKBPSLNJNKFVAC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100320

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-100320
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart