ENA371177619 Enamine

1-methyl-5-(thiophen-2-yl)-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD10036229
  • Cas Number : 1177332-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106754
Availability details for
Quantity
  • Availability
  • Add to Cart