ENA371177621 Enamine

2-(chloromethyl)-6-ethylpyrimidin-4-ol

  • MDL number MFCD22375283
  • Cas Number : 344294-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106756
Availability details for
Quantity
  • Availability
  • Add to Cart