ENA371177623 Enamine

3-tert-butyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12462889
  • Cas Number : 1152890-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106758
Availability details for
Quantity
  • Availability
  • Add to Cart