ENA371177626 Enamine

3-bromo-4-(4-bromophenyl)-4-oxobutanoic acid

  • MDL number MFCD22375215
  • Cas Number : 35158-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101177

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101177
Availability details for
Quantity
  • Availability
  • Add to Cart