ENA371177630 Enamine

trans-(4-aminocyclohexyl)methanol

  • MDL number MFCD12407131
  • Cas Number : 1467-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106767

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106767
Availability details for
Quantity
  • Availability
  • Add to Cart