ENA371177632 Enamine

3,6-dimethylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD01693052
  • Cas Number : 1431-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106770
Availability details for
Quantity
  • Availability
  • Add to Cart