ENA371177633 Enamine

1-(2-fluorophenyl)-3,5-dimethyl-1H-pyrazol-4-amine

  • MDL number MFCD11916117
  • Cas Number : 1153040-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106771

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106771
Availability details for
Quantity
  • Availability
  • Add to Cart