ENA371177635 Enamine

3-amino-1-(furan-2-yl)butan-1-ol

  • MDL number MFCD22375288
  • Cas Number : 1384533-85-0
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-1-(2-furyl)-1-butanol
Inchi Code 1S/C8H13NO2/c1-6(9)5-7(10)8-3-2-4-11-8/h2-4,6-7,10H,5,9H2,1H3
Inchi Key WAFBZZZRVPRDGT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106775
Availability details for
Quantity
  • Availability
  • Add to Cart