ENA371177638 Enamine

(4-aminocyclohexanecarbonyl)urea hydrochloride

  • MDL number MFCD22375291
  • Cas Number : 1384563-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106784
Availability details for
Quantity
  • Availability
  • Add to Cart