ENA371177643 Enamine

2-(3-chlorophenyl)-1-methyl-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD07377679
  • Cas Number : 1394041-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106793
Availability details for
Quantity
  • Availability
  • Add to Cart