ENA371177646 Enamine

1-(5-methyl-1,3,4-oxadiazol-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22375293
  • Cas Number : 1384662-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106798
Availability details for
Quantity
  • Availability
  • Add to Cart