ENA371177647 Enamine

1-methanesulfonyl-N-methylpiperidin-4-amine hydrochloride

  • MDL number MFCD22375294
  • Cas Number : 1384565-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106801
Availability details for
Quantity
  • Availability
  • Add to Cart