ENA371177648 Enamine

5-chloro-2-(3-chlorophenoxy)benzoic acid

  • MDL number MFCD18580142
  • Cas Number : 69199-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106802
Availability details for
Quantity
  • Availability
  • Add to Cart