ENA371177650 Enamine

(5-chloro-2-fluorophenyl)methanesulfonyl chloride

  • MDL number MFCD11976099
  • Cas Number : 1314961-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45...47
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104243

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104243
Availability details for
Quantity
  • Availability
  • Add to Cart