ENA371177657 Enamine

5-(bromomethyl)-2-fluorobenzonitrile

  • MDL number MFCD03094341
  • Cas Number : 180302-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106813

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-106813
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart