ENA371177671 Enamine

5-bromo-4-chlorothiophene-2-sulfonamide

  • MDL number MFCD00177162
  • Cas Number : 77893-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106870
Availability details for
Quantity
  • Availability
  • Add to Cart