ENA371177678 Enamine

[4-(methylamino)cyclohexyl]methanol

  • MDL number MFCD11520458
  • Cas Number : 1260831-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106883
Availability details for
Quantity
  • Availability
  • Add to Cart