ENA371177680 Enamine

N-ethyl-N-methylformamide

  • MDL number MFCD21103047
  • Cas Number : 28860-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106885

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106885
Availability details for
Quantity
  • Availability
  • Add to Cart