ENA371177683 Enamine

4-(4-iodophenoxymethyl)pyridine

  • MDL number MFCD14648758
  • Cas Number : 944278-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106889

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106889
Availability details for
Quantity
  • Availability
  • Add to Cart